Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101947 Supplier Invitrogen™ Supplier No. PA5101947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UBAC1.

UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBAC1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UBAC1
Gene Accession No. Q9BSL1
Gene Alias 1110033G07Rik; AA407978; E3 ubiquitin-protein ligase subunit KPC2; GBDR1; Gdbr1; glialblastoma cell differentiation-related protein 1; Kip1 ubiquitination-promoting complex protein 2; KPC2; putative glialblastoma cell differentiation-related; putative glialblastoma cell differentiation-related protein; RP11-432J22.3; testicular secretory protein Li 64; UBA domain containing 1; UBA domain-containing protein 1; Ubac1; Ubadc1; ubiquitin associated domain containing 1; ubiquitin-associated domain-containing protein 1
Gene Symbols UBAC1
Host Species Rabbit
Immunogen A synthesized peptide derived from human UBAC1(Accession Q9BSL1), corresponding to amino acid residues K94-N144.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10422
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.