Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBAC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553063
Description
Immunogen sequence: YDSKMFQVHQ VLCIPSWMAK FFSWTLEPIF SSSEPTSEAR IGMGATLDIQ RQQRMELLDR QLMFSQFAQG RRQRQQQGGM INWNRLFPPL RQRQNVNYQG GRQSEPAAPP LEVSEEQVAR LMEMGFSRGD ALEALRASNN DLN Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

Specifications
UBAC2 | |
Polyclonal | |
Unconjugated | |
Ubac2 | |
1190008A14Rik; AW554959; PHGDHL1; phosphoglycerate dehydrogenase like 1; phosphoglycerate dehydrogenase-like protein 1; PSEC0110; RP11-178C10.1; UBA domain containing 2; UBA domain-containing protein 2; Ubac2; ubiquitin associated domain containing 2; ubiquitin-associated domain-containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
337867, 361094, 68889 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NBM4, Q8R1K1 | |
Ubac2 | |
Recombinant protein corresponding to Human UBAC2. Recombinant protein control fragment (Product #RP-90960). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction