Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554085 Supplier Invitrogen™ Supplier No. PA554085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FHGTFSYLDD VPFKTGDKFK TPAKVGLPIG FSLPDCLQVV REVQYDFSLE KKTIEWAEEI KKIEEAEREA ECKIAEAEAK VNSKSGPEGD SKMSFSKTHS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBAP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBAP1
Gene Accession No. Q9NZ09
Gene Alias 2700092A01Rik; NAG20; nasopharyngeal carcinoma-associated gene 20 protein; UAP; Ubap; Ubap1; UBAP-1; ubiquitin associated protein 1; ubiquitin-associated protein 1
Gene Symbols UBAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBAP1. Recombinant protein control fragment (Product #RP-93033).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.