Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $530.00
Specifications
Antigen | UBAP2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119494
|
Invitrogen™
PA5119494 |
200 μL |
Each for $530.00
|
|
|||||
PIPA5121513
|
Invitrogen™
PA5121513 |
20 μL |
Each for $209.50
|
|
|||||
Description
UBAP2 Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene contains a UBA (ubiquitin associated) domain, which is characteristic of proteins that function in the ubiquitination pathway. This gene may show increased expression in the adrenal gland and lymphatic tissues. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Sep 2013].Specifications
UBAP2 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
UBAP2 | |
1190005K07Rik; AA408600; AD-012 protein; AU045235; KIAA1491; Lig1; mKIAA1491; mLig-1; protein lingerer homolog 1; ubap2; UBAP-2; ubiquitin associated protein 2; ubiquitin-associated protein 2 | |
UBAP2 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q5T6F2 | |
55833 | |
Recombinant protein Ubiquitin Associated Protein 2. The antigen corresponds to amino acid range 1-104 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title