Learn More
UBAP2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304627A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene contains a UBA (ubiquitin associated) domain, which is characteristic of proteins that function in the ubiquitination pathway. This gene may show increased expression in the adrenal gland and lymphatic tissues. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Sep 2013]Specifications
UBAP2 | |
Polyclonal | |
Unconjugated | |
UBAP2 | |
Ubiquitin-associated protein 2, UBAP-2, UBAP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55833 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q5T6F2 | |
UBAP2 | |
Between 600 and 650 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.