Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBAP2L Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100751JF646
Description
UBAP2L Polyclonal antibody specifically detects UBAP2L in Human samples. It is validated for Western BlotSpecifications
UBAP2L | |
Polyclonal | |
Western Blot | |
FLJ42300, KIAA0144NICE-4, NICE4, Protein NICE-4, ubiquitin associated protein 2-like, ubiquitin-associated protein 2-like | |
The immunogen recognized by this antibody maps to a region between residue 1025 and the C-terminus (residue 1087) of human Ubiquitin Associated Protein 2-Like using the numbering given in entry AAH03170.1 (GeneID 9898). | |
0.1 mL | |
Cell Biology | |
9898 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction