Learn More
UBASH3A Mouse anti-Human, Clone: OTI5B4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809272

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TULA (T cell ubiquitin ligand, UBASH3A, ubiquitin associated and SH3 domain containing A), STS-2 or CLIP4, is a 661 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one SH3 domain and one UBA domain. Expressed at high levels in thymus, bone marrow, spleen and peripheral blood leukocytes, TULA exists as either a homodimer or a homo-oligomer that interferes with the degradation of receptor-type tyrosine kinases and promotes the accumulation of activated receptors on the cell surface. Additionally, TULA is part of an EGFR- and Cbl-containing complex that interacts with ubiquitinated proteins. The gene encoding TULA, which maps to human chromosome 21, may be involved in the pathogenesis of type I diabetes. Multiple isoforms of TULA are produced due to alternative splicing events.Specifications
UBASH3A | |
Monoclonal | |
Unconjugated | |
UBASH3A | |
CLIP4, STS-2, TULA, TULA-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53347 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5B4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P57075 | |
UBASH3A | |
Human recombit protein fragment corresponding to amino acids 288-549 of human UBASH3A produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.