Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBC13 Recombinant Rabbit Monoclonal Antibody (JG38-68)
GREENER_CHOICE

Catalog No. PIMA534786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534786 Supplier Invitrogen™ Supplier No. MA534786
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Daudi, SH-SY-5Y, mouse spleen tissue lysate, rat spleen tissue lysate, human colon tissue, human prostate cancer tissue, mouse testis tissue.

The UBE2V1-UBE2N and UBE2V2-UBE2N heterodimers catalyze the synthesis of non-canonical 'Lys-63'-linked polyubiquitin chains. This type of polyubiquitination does not lead to protein degradation by the proteasome. Mediates transcriptional activation of target genes. Plays a role in the control of progress through the cell cycle and differentiation. Plays a role in the error-free DNA repair pathway and contributes to the survival of cells after DNA damage. Acts together with the E3 ligases, HLTF and SHPRH, in the 'Lys-63'-linked poly-ubiquitination of PCNA upon genotoxic stress, which is required for DNA repair. Appears to act together with E3 ligase RNF5 in the 'Lys-63'-linked polyubiquitination of JKAMP thereby regulating JKAMP function by decreasing its association with components of the proteasome and ERAD. Promotes TRIM5 capsid-specific restriction activity and the UBE2V1-UBE2N heterodimer acts in concert with TRIM5 to generate 'Lys-63'-linked polyubiquitin chains which activate the MAP3K7/TAK1 complex which in turn results in the induction and expression of NF-kappa-B and MAPK-responsive inflammatory genes. Together with RNF135 and UB2V1, catalyzes the viral RNA-dependent 'Lys-63'-linked polyubiquitination of RIG-I/DDX58 to activate the downstream signaling pathway that leads to interferon beta production. UBE2V1-UBE2N together with TRAF3IP2 E3 ubiquitin ligase mediate 'Lys-63'-linked polyubiquitination of TRAF6, a component of IL17A-mediated signaling pathway. (Uniprot).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBC13
Applications Western Blot
Classification Recombinant Monoclonal
Clone JG38-68
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene UBE2N
Gene Accession No. P61088
Gene Alias 1500026J17Rik; AL022654; BB101821; bendless protein; bendless-like ubiquitin conjugating enzyme; bendless-like ubiquitin-conjugating enzyme; Blu; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; HEL-S-71; MGC131857; MGC8489; UBC13; UbcH13; UbcH-ben; UBCHBEN; UBC13; UBE2N; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2 N; ubiquitin conjugating enzyme E2N; ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); ubiquitin-protein ligase N; yeast UBC13 homolog
Gene Symbols UBE2N
Host Species Rabbit
Immunogen Recombinant protein within Human Ube2N aa 1-93.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7334
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.