Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBC13Goat anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991566
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1566 Supplier Rockland Immunochemicals Supplier No. 600101342
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against human UBC13 protein. A BLAST analysis was used to suggest cross reactivity with UBC13 protein from a broad range of sources including human, dog, bovine, Xenopus laevis, Xenopus tropicalis, , zebrafish, rat, mouse, chimpanzee, macaque and chicken based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

Anti-Ubc13 is a monoclonal antibody specific to the ∼19 kDa Ubc13 protein. Ubc13 is a ubiquitin conjugating enzyme that is complexed with a ubiquitin activating enzyme (E1), Mms2. The association of Mms2 to Ubc13 forms an active heterodimer that is involved in error-free post replicative DNA repair in yeast. Upon complexing with another conjugating enzyme variant, Uev1a, Ubc13 participates in the regulation of NF-kB signal transduction pathway in humans.

Specifications

Antigen UBC13
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene UBE2N
Gene Accession No. P61088
Gene Alias 1500026J17Rik; AL022654; BB101821; bendless protein; bendless-like ubiquitin conjugating enzyme; Bendless-like ubiquitin-conjugating enzyme; BLU; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; HEL-S-71; MGC131857; MGC8489; Ubc13; UbcH13; UbcH-ben; UBCHBEN; UBC13; Ube2n; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2 N; ubiquitin conjugating enzyme E2N; Ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); Ubiquitin-protein ligase N; yeast UBC13 homolog
Gene Symbols UBE2N
Host Species Goat
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7334
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.