Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566358 Supplier Invitrogen™ Supplier No. PA566358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKTKVPSNDNS SDLLYDDLYD DD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Protein kinase CK2 is a ubiquitous and pleiotropic Ser/Thr protein kinase involved in cell growth and transformation. This gene encodes a protein similar to the E2 ubiquitin conjugating enzyme UBC3/CDC34. Studies suggest that CK2-dependent phosphorylation of this ubiquitin-conjugating enzyme functions by regulating beta-TrCP substrate recognition and induces its interaction with beta-TrCP, enhancing beta-catenin degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBC3B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2R2
Gene Accession No. Q712K3
Gene Alias 1200003M11Rik; CDC34B; cell division cycle 34 homolog; E2 ubiquitin-conjugating enzyme R2; E2-CDC34B; LOC689226; similar to ubiquitin-conjugating enzyme E2R 2; UBC3B; Ube2r2; Ubiquitin carrier protein R2; ubiquitin conjugating enzyme E2 R2; ubiquitin conjugating enzyme E2R 2; ubiquitin-conjugating enzyme E2 R2; ubiquitin-conjugating enzyme E2-32k; ubiquitin-conjugating enzyme E2-CDC34B; ubiquitin-conjugating enzyme E2R 2; ubiquitin-conjugating enzyme UBC3B; ubiquitin-protein ligase R2
Gene Symbols UBE2R2
Host Species Rabbit
Immunogen Recombinant Human UBC3B. Recombinant protein control fragment (Product #RP-105163).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.