Learn More
Invitrogen™ UBC9 Monoclonal Antibody (1B10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515916
Description
MA5-15916 targets UBE2I in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-15916 immunogen is purified recombinant fragment of human UBE2I expressed in E. Coli. . MA5-15916 detects UBE2I which has a predicted molecular weight of approximately 18kDa.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
UBC9 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P63279, P63280 | |
UBE2I | |
Purified recombinant fragment of human UBE2I expressed in E. Coli. | |
RUO | |
22196, 7329 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1B10 | |
Unconjugated | |
UBE2I | |
5830467E05Rik; C358B7.1; F830028O17Rik; mUBC9; OTTHUMP00000198131; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; UBC9; UbcE2A; Ubce2i; Ubce9; Ube2i; Ubiquitin carrier protein 9; ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2 I; ubiquitin conjugating enzyme E2I; Ubiquitin conjugating enzyme E2I (homologous to yeast UBC9); Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; ubiquitin-protein ligase I | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.