Learn More
Invitrogen™ UBC9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585564
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: 293T, A431, HeLa, HepG2, mouse testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| UBC9 | |
| Polyclonal | |
| Unconjugated | |
| UBE2I | |
| 5830467E05Rik; C358B7.1; F830028O17Rik; mUBC9; OTTHUMP00000198131; p18; RING-type E3 SUMO transferase UBC9; SUMO-1-protein ligase; SUMO-conjugating enzyme UBC9; SUMO-protein ligase; UBC9; UbcE2A; Ubce2i; Ubce9; Ube2i; Ubiquitin carrier protein 9; ubiquitin carrier protein I; ubiquitin conjugating enzyme 9; ubiquitin conjugating enzyme E2 I; ubiquitin conjugating enzyme E2I; Ubiquitin conjugating enzyme E2I (homologous to yeast UBC9); Ubiquitin-conjugating enzyme E2 I; ubiquitin-conjugating enzyme E2I; ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9); ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast); ubiquitin-conjugating enzyme UbcE2A; ubiquitin-like protein SUMO-1 conjugating enzyme; ubiquitin-protein ligase E2I; ubiquitin-protein ligase I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22196, 7329 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P63279, P63280 | |
| UBE2I | |
| Recombinant protein encompassing a sequence within the center region of human UBE2I. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.