Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH10/UBE2C Antibody (CPTC-UBE2C-1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308829IR
Description
UbcH10/UBE2C Monoclonal antibody specifically detects UbcH10/UBE2C in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
UbcH10/UBE2C | |
Monoclonal | |
DyLight 755 | |
cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C | |
Recombinant human full-length UbcH10/UBE2C protein (Uniprot: O00762) | |
100 μg | |
Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators | |
11065 | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA, Immunohistochemistry (Paraffin) | |
CPTC-UBE2C-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction