Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH10/UBE2C Antibody (SR1319), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | UbcH10/UBE2C |
---|---|
Clone | SR1319 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
UbcH10/UBE2C Monoclonal antibody specifically detects UbcH10/UBE2C in Human samples. It is validated for Western BlotSpecifications
UbcH10/UBE2C | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C | |
A synthesized peptide derived from human UbcH10/UBE2C (Uniprot #: O00762) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1319 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
11065 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title