Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH2/UBE2H Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300048AF488
Description
UbcH2/UBE2H Polyclonal specifically detects UbcH2/UBE2H in Human samples. It is validated for Western Blot, ELISA.Specifications
UbcH2/UBE2H | |
Polyclonal | |
Western Blot, ELISA | |
EC 6.3.2.19, UBC8E2-20K, UBCH, UBCH2, Ubiquitin carrier protein H, ubiquitin-conjugating enzyme E2 H, Ubiquitin-conjugating enzyme E2-20K, ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8), ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast), Ubiquitin-protein ligase H | |
Produced in rabbits immunized with purified, recombinant UbcH2/UBE2H (P62256; Met1-Leu183). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
7328 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction