Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH5a/UBE2D1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299796G
Description
UbcH5a/UBE2D1 Polyclonal specifically detects UbcH5a/UBE2D1 in Human samples. It is validated for Western Blot, ELISA.Specifications
UbcH5a/UBE2D1 | |
Polyclonal | |
Western Blot, ELISA | |
E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1 | |
Produced in rabbits immunized with purified, recombinant UbcH5a/UBE2D1 (P51668; Ala2-Met147). | |
0.1 mL | |
Cancer, Hypoxia | |
7321 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction