Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH5a/UBE2D1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155033
Description
UbcH5a/UBE2D1 Polyclonal antibody specifically detects UbcH5a/UBE2D1 in Human, Rat samples. It is validated for Western Blot.Specifications
UbcH5a/UBE2D1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1 | |
Rabbit | |
16 kDa | |
100 μL | |
Cancer, Hypoxia | |
7321 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
P51668 | |
UBE2D1 | |
Synthetic peptides corresponding to UBE2D1(ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast)) The peptide sequence was selected from the middle region of UBE2D1. Peptide sequence SQWSPALTVSKVLLSICSLLCDPNPDDPLVPDIAQIYKSDKEKYNRHARE. | |
Protein A purified | |
RUO | |
Primary | |
Guinea pig: 79%. | |
Human, Rat, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction