Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH5b/UBE2D2 Antibody (OTI2C2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274732JF646
Description
UbcH5b/UBE2D2 Monoclonal specifically detects UbcH5b/UBE2D2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
UbcH5b/UBE2D2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UBE2D2 | |
Full length recombinant protein of human UbcH5b/UBE2D2 (NP_003330) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI2C2 | |
Western Blot, Immunohistochemistry | |
E2(17)KB2, EC 6.3.2.19, PUBC1, UBC4/5, UBC4ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5), UBC5B, UBCH4, UbcH5B, Ubiquitin carrier protein D2, ubiquitin-conjugating enzyme E2 D2, Ubiquitin-conjugating enzyme E2(17)KB 2, Ubiquitin-conjugating enzyme E2-17 kDa 2, ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D2 | |
Mouse | |
Affinity Purified | |
Cancer, Hypoxia | |
7322.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction