Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH6/UBE2E1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178983AF350
Description
UbcH6/UBE2E1 Polyclonal specifically detects UbcH6/UBE2E1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UbcH6/UBE2E1 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 6.3.2.19, UbcH6, Ubiquitin carrier protein E1, ubiquitin-conjugating enzyme E2 E1, ubiquitin-conjugating enzyme E2E 1 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2E 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E1 | |
Rabbit | |
Affinity Purified | |
Epigenetics | |
7324.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
UBE2E1 | |
A synthetic peptide made to an N-terminal portion of the human UBCH6 protein (between residues 1-50) [UniProt P51965] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction