Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH7/UBE2L3 Antibody (SR1268), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | UbcH7/UBE2L3 |
---|---|
Clone | SR1268 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
UbcH7/UBE2L3 Monoclonal antibody specifically detects UbcH7/UBE2L3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
UbcH7/UBE2L3 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
E2-F1, EC 6.3.2.19, L-UBC, UBCE7, UbcH7, UBCH7UbcM4, Ubiquitin carrier protein L3, ubiquitin-conjugating enzyme E2 L3, Ubiquitin-conjugating enzyme E2-F1, ubiquitin-conjugating enzyme E2L 3, ubiquitin-conjugating enzyme UBCH7, Ubiquitin-protein ligase L3 | |
A synthesized peptide derived from human UbcH7/UBE2L3 (Uniprot #: P68036) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1268 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cancer, DNA Repair, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7332 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title