Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH8/Ube2L6 Antibody (140), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290137F
Description
UbcH8/Ube2L6 Monoclonal specifically detects UbcH8/Ube2L6 in Human samples. It is validated for ELISA.Specifications
UbcH8/Ube2L6 | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
140 | |
ELISA | |
EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human UbcH8/Ube2L6 (O14933-1; Met1-Ser153). | |
0.1 mL | |
Stem Cell Markers | |
9246 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction