Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UbcH8/Ube2L6 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300032PE
Description
UbcH8/Ube2L6 Polyclonal antibody specifically detects UbcH8/Ube2L6 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
UbcH8/Ube2L6 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
EC 6.3.2.19, retinoic acid induced gene B protein, Retinoic acid-induced gene B protein, RIG-B, UbcH8, UBCH8MGC40331, Ubiquitin carrier protein L6, ubiquitin/ISG15-conjugating enzyme E2 L6, ubiquitin-conjugating enzyme E2L 6, Ubiquitin-protein ligase L6 | |
Produced in rabbits immunized with purified, recombinant Human UbcH8/Ube2L6 (Accession#: O14933-1; Met1-Ser153) | |
0.1 mL | |
Stem Cell Markers | |
9246 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction