Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBD Recombinant Rabbit Monoclonal Antibody (10A6T5)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA535744

Catalog No. PIMA535744


Only null left
Add to Cart

Description

Description

Immunogen sequence: KEKTIHLTLK VVKPSDEELP LFLVESGDEA KRHLLQVRRS SSVAQVKAMI ETKTGIIPET QIVTCNGKRL EDGKMMADYG I.

UBD qualifies as a marker for an interferon response in hepatocellular carcinoma and colon carcinoma but is not significantly overexpressed in cancers lacking a proinflammatory environment. Immunohistochemical studies demonstrated increased UBD expression in HIV-associated nephropathy and in autosomal dominant polycystic kidney disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UBD
Recombinant Monoclonal
2.8 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O15205, P63072, Q921A3
UBD
A synthetic peptide corresponding to a sequence within amino acids 70-150 of human FAT10/UBD (O15205).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot, Immunocytochemistry
10A6T5
Unconjugated
UBD
Diubiquitin; FAT10; GABBR1; Ubd; UBD 3; UBD-3; Ubiquitin D; ubiquitin-like protein FAT10
Rabbit
Affinity chromatography
RUO
10537, 24108, 29168
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.