Learn More
Invitrogen™ UBD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596345
Description
Immunogen sequence: MAPNASCLCV HVRSEEWDLM TFDANPYDSV KKIKEHVRSK TKVPVQDQVL LLGSKILKPR RSLSSYGIDK EKTIHLTLKV VKPSDEELPL FLVESGDEAK RHLLQVRRSS SVAQVKAMIE TKTGIIPETQ IVTCNGKRLE DGKMMADYGI RKGNLLFLAC YCIGG; Positive Samples: Mouse liver; Cellular Location: Cytoplasm, Nucleus.
UBD qualifies as a marker for an interferon response in hepatocellular carcinoma and colon carcinoma but is not significantly overexpressed in cancers lacking a proinflammatory environment. Immunohistochemical studies demonstrated increased UBD expression in HIV-associated nephropathy and in autosomal dominant polycystic kidney disease.
Specifications
UBD | |
Polyclonal | |
Unconjugated | |
UBD | |
Diubiquitin; FAT10; GABBR1; Ubd; UBD 3; UBD-3; Ubiquitin D; ubiquitin-like protein FAT10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10537, 24108 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.73 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15205, P63072 | |
UBD | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human FAT10/FAT10/UBD (NP_0063892). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.