Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE1 Monoclonal Antibody (05)
GREENER_CHOICE

Catalog No. PIMA529631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529631 Supplier Invitrogen™ Supplier No. MA529631
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human UBE1/UBA1.

UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% trehalose and no preservative
Gene UBA1
Gene Accession No. P22314
Gene Alias A1S9; A1S9 protein; A1S9T; A1S9T and BN75 temperature sensitivity complementing; A1ST; AMCX1; CFAP124; EMBL:AAH85791.1, ECO:0000312; GXP1; POC20; POC20 centriolar protein homolog; Protein A1S9; RGD:1359327}; Sbx; SMAX2; testicular secretory protein Li 63; Uba1; uba1 {ECO:0000312; UBA1, ubiquitin-activating enzyme E1 homolog A; UBA1A; UBE1; Ube-1; Ube1ax; Ube1x; ubiquitin like modifier activating enzyme 1; Ubiquitin-activating enzyme E1; ubiquitin-activating enzyme E1 {ECO:0000250; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; ubiquitin-like modifier-activating enzyme 1; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250; Ubiquitin-like modifier-activating enzyme 1 X; UniProtKB:Q02053}
Gene Symbols UBA1
Host Species Mouse
Immunogen Recombinant Human UBE1/UBA1 protein (Ser2-Arg1058).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.