Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588493 Supplier Invitrogen™ Supplier No. PA588493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KISFKSLVAS LAEPDFVVTD FAKFSRPAQL HIGFQALHQF CAQHGRPPRP RNEEDAAELV ALAQAVNARA LPAVQQNNLD EDLIRKLAYV AAGDLAPINA FIGGLAAQEV MKACSGKFMP IMQWLYFDAL ECLPEDKEVL TEDKCLQRQN RYDGQVAVFG SDLQEKLGKQ KYFLVGAGAI GCELLKNFAM IGLGCGEGGE IIVTDMDTIE KSNLNRQFLF RPWDVTKLKS DTAAAAVRQM NPHIRVTSHQ NRVGPDTERI YDDDFFQNLD GVANALDNVD ARMYMDRRCV YYRKPLLESG T; Positive Samples: U-251MG, Rat spleen; Cellular Location: Cytoplasm, Mitochondrion, Nucleus, Nucleus.

UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UBA1
Gene Accession No. P22314, Q02053, Q5U300
Gene Alias A1S9; A1S9 protein; A1S9T; A1S9T and BN75 temperature sensitivity complementing; A1ST; AMCX1; CFAP124; EMBL:AAH85791.1, ECO:0000312; GXP1; POC20; POC20 centriolar protein homolog; Protein A1S9; RGD:1359327}; Sbx; SMAX2; testicular secretory protein Li 63; Uba1; uba1 {ECO:0000312; UBA1, ubiquitin-activating enzyme E1 homolog A; UBA1A; UBE1; Ube-1; Ube1ax; Ube1x; ubiquitin like modifier activating enzyme 1; Ubiquitin-activating enzyme E1; ubiquitin-activating enzyme E1 {ECO:0000250; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; ubiquitin-like modifier-activating enzyme 1; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250; Ubiquitin-like modifier-activating enzyme 1 X; UniProtKB:Q02053}
Gene Symbols UBA1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human UBA1 (NP_6950121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22201, 314432, 7317
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.