Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA511959
Description
This antibody is predicted to react with bovine, rat and rabbit based on sequence homology.
UBE1 catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11. 23. Alternative splicing results in 2 transcript variants encoding the same protein, but with different 5' UTR.
Specifications
UBE1 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
P22314, Q02053 | |
UBA1 | |
KLH conjugated synthetic peptide between 1026-1058 amino acids from the C-terminal region of human E1 Ubiquitin (UBE1). | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
UBA1 | |
A1S9; A1S9 protein; A1S9T; A1S9T and BN75 temperature sensitivity complementing; A1ST; AMCX1; CFAP124; EMBL:AAH85791.1, ECO:0000312; GXP1; POC20; POC20 centriolar protein homolog; Protein A1S9; RGD:1359327}; Sbx; SMAX2; testicular secretory protein Li 63; Uba1; uba1 {ECO:0000312; UBA1, ubiquitin-activating enzyme E1 homolog A; UBA1A; UBE1; Ube-1; Ube1ax; Ube1x; ubiquitin like modifier activating enzyme 1; Ubiquitin-activating enzyme E1; ubiquitin-activating enzyme E1 {ECO:0000250; Ubiquitin-activating enzyme E1 X; ubiquitin-activating enzyme E1, Chr X; ubiquitin-like modifier activating enzyme 1; ubiquitin-like modifier-activating enzyme 1; ubiquitin-like modifier-activating enzyme 1 {ECO:0000250; Ubiquitin-like modifier-activating enzyme 1 X; UniProtKB:Q02053} | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
22201, 7317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction