Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596707 Supplier Invitrogen™ Supplier No. PA596707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEGSEPVAAH QGEEASCSSW GTGSTNKNLP IMSTASVEID DALYSRQRYV LGDTAMQKMA KSHVFLSGMG GLGLEIAKNL VLAGIKAVTI HDTEKCQAWD LGTNFFLSED DVVNKRNRAE AVLKHIAELN PYVHVTSSSV PFNETTDLSF LDKYQCVVLT EMKLPLQKKI NDFCRSQCPP IKFISADVHG IWSRLFCDFG DEFEVLDTTG EEPKEIFISN ITQANPGIVT CLENHPHKLE TGQFLTFREI NGMTGLNGSI; Positive Samples: U-251MG, SKOV3, 293T, Mouse brain.

UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins.UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility.UBA6 is highly expressed in human monocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE1L2
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBA6
Gene Accession No. A0AVT1, Q8C7R4
Gene Alias 4930542H01; 5730469D23Rik; AU021846; AW124799; E1-L2; Monocyte protein 4; MOP4; MOP-4; RGD1308324; UBA6; UBA6, ubiquitin-activating enzyme E1; Ube1l2; ubiquitin like modifier activating enzyme 6; ubiquitin-activating enzyme 6; ubiquitin-activating enzyme E1-like 2; ubiquitin-activating enzyme E1-like protein 2; ubiquitin-like modifier activating enzyme 6; ubiquitin-like modifier-activating enzyme 6
Gene Symbols UBA6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human UBA6 (NP_0606974).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231380, 55236
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.