Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527186 Supplier Invitrogen™ Supplier No. PA527186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (100%), Rat (93%), Zebrafish (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Multiple alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene UBE2A
Gene Accession No. P49459
Gene Alias E2 ubiquitin-conjugating enzyme A; hhr6a; HR6A; Mhr6a; mp:zf637-2-000771; MRXS30; MRXSN; RAD6 homolog A; Rad6a; si:bz46j2.4; UBC2; ube2a; ube2a.L; Ubiquitin carrier protein A; ubiquitin conjugating enzyme E2 A; ubiquitin conjugating enzyme E2 A L homeolog; ubiquitin conjugating enzyme E2 A L homeolog; ubiquitin conjugating enzyme E2A L homeolog; ubiquitin conjugating enzyme E2A; ubiquitin-conjugating enzyme E2 A; ubiquitin-conjugating enzyme E2A; ubiquitin-conjugating enzyme E2A (RAD6 homolog); ubiquitin-conjugating enzyme E2A, RAD6 homolog; ubiquitin-conjugating enzyme HR6A; ubiquitin-protein ligase A; wu:fa01h11; XELAEV_18038882mg
Gene Symbols UBE2A
Host Species Rabbit
Immunogen Full length human UBE2A Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.