Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2A, Rabbit anti-Human, Mouse, Polyclonal, Millipore Sigma™

Catalog No. ABS220325UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS220325UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS220325UG Supplier MilliporeSigma Supplier No. ABS220325UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-UBE2A Antibody, Cat. No. ABS2203, is a rabbit polyclonal antibody that detects UBE2A and has been tested for use Immunohistochemistry (Paraffin) and Western Blotting.

Ubiquitin-conjugating enzyme E2 A (UniProt: P49459; also known as EC 2.3.2.23; E2 ubiquitin-conjugating enzyme A, RAD6 homolog A, HR6A, hHR6A, Ubiquitin carrier protein A, Ubiquitin-protein ligase A) is encoded by the UBE2A (also known as RAD6A) gene (GeneID: 7319) in human. UBE2A belongs to the ubiquitin-conjugating enzyme family and accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In association with the E3 enzyme BRE1, it plays a role in transcription regulation by catalyzing the mono-ubiquitination of histone H2B at Lys-120 that provides a specific tag for epigenetic transcriptional activation, elongation by RNA polymerase II, and telomeric silencing. It is also a prerequisite for H3K4me and H3K79me formation. Defects in UBE2A gene have been linked to mental retardation, X-linked, syndromic, Nascimento-type mental retardation that is characterized by significantly below average general intellectual functioning associated with impairments in adaptive behavior.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:500 dilution from a representative lot detected UBE2A in mouse kidney tissue lysate.Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected UBE2A in human pancreas, human cervical cancer, and human uterus tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ubiquitin-conjugating enzyme E2A;EC 2.3.2.23;E2 ubiquitin-conjugating enzyme A;RAD6 homolog A;HR6A;hHR6A;Ubiquitin carrier protein A;Ubiquitin-protein ligase A
Gene Symbols UBE2A;RAD6A
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Ubiquitin-conjugating enzyme E2 A (UBE2A).
Molecular Weight of Antigen ∽17 kDa observed; 17.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Ubiquitin-conjugating enzyme E2 A (UBE2A) in HeLa cells and Mouse Kidney.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.