Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | UBE2A |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6858
![]() ![]() |
Proteintech Group Inc
110801AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
LNPEP, also known as insulin-responsive aminopeptidase (IRAP), was initially identified as an abundant protein that is associated with specialized endosomes termed GLUT4 storage vesicles (GSVs) in adipocytes. The luminal domain of LNPEP contains an aminopeptidase activity that is involved in peptide hormone processing and antigenic peptide processing. LNPEP is also a key regulator of GLUT4 trafficking by controlling the sorting of GLUT4 from endosomes to the GSVs.Specifications
UBE2A | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P49459, Q9Z255 | |
22209, 298317, 7319 | |
UBE2A Fusion Protein Ag1567 | |
Primary | |
-20°C | |
UBE2A |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HHR6A, HR6A, RAD6 homolog A, RAD6A, UBC2, UBE2A, Ubiquitin carrier protein A, Ubiquitin protein ligase A | |
UBE2A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title