Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110674
Description
Immunogen sequence: LMRDFKRLQE DPPVGVSGAP SENNIMQWNA VIFGPEGTPF EDGTFKLVIE FSEEYPNKPP TVRFLSKMFH PNVYADGSIC LDILQNRWSP TYDVSSILTS IQSLLDEPNP NSPANSQAAQ.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2B is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.
Specifications
| UBE2B | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P63146 | |
| Ube2b | |
| Recombinant Protein corresonding to Human UBE2B. Recombinant protein control fragment (Product #RP-105607). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| Unconjugated | |
| Ube2b | |
| 2610301N02Rik; E2 protein; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; E2-14k; E2-17kDa; HHR6B; HR6B; mHR6B; RAD6 homolog B; Rad6b; UBC2; Ube2b; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2 B; ubiquitin conjugating enzyme E2B; ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-protein ligase B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7320 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction