Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2B Recombinant Rabbit Monoclonal Antibody (JE56-12)
GREENER_CHOICE

Catalog No. PIMA542490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542490 Supplier Invitrogen™ Supplier No. MA542490
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: A431 cell lysate, THP-1 cell lysate, HepG2 cell lysate, zebrafish tissue lysates, human breast carcinoma tissue, human fetal skeletal muscle tissue. Subcellular Location: Nucleus, Cell membrane.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2B is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE56-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Ube2b
Gene Accession No. P63146
Gene Alias 2610301N02Rik; E2 protein; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; E2-14k; E2-17kDa; HHR6B; HR6B; mHR6B; RAD6 homolog B; Rad6b; UBC2; Ube2b; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2 B; ubiquitin conjugating enzyme E2B; ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-protein ligase B
Gene Symbols Ube2b
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal Human Ube2B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 437020, 7320
Target Species Human, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.