Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117019 Supplier Invitrogen™ Supplier No. PA5117019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 22Rv1, THP-1, U-87MG, SW620, SKOV3 Immunogen sequence: MASQNRDPAA TSVAAARKGA EPSGGAARGP VGKRLQQELM TLMMSGDKGI SAFPESDNLF KWVGTIHGAA GTVYEDLRYK LSLEFPSGYP YNAPTVKFLT PCYHPNVDTQ GNICLDILKE KWSALYDVRT ILLSIQSLLG EPNIDSPLNT HAAELWKNPT AFKKYLQETY SKQVTSQEP.

This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is required for the destruction of mitotic cyclins and for cell cycle progression, and may be involved in cancer progression. Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-11'- and 'Lys-48'-linked polyubiquitination. Acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated ubiquitin ligase that controls progression through mitosis. Acts by initiating 'Lys-11'-linked polyubiquitin chains on APC/C substrates, leading to the degradation of APC/C substrates by the proteasome and promoting mitotic exit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2C
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene UBE2C
Gene Accession No. O00762, Q9D1C1
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme C; 1110015A16Rik; cyclin-selective ubiquitin carrier protein; D2Ertd695e; dJ447F3.2; E2 ubiquitin-conjugating enzyme C; mitotic-specific ubiquitin-conjugating enzyme; RP3-447F3.1; ubcH10; UBE2C; Ubiquitin carrier protein C; ubiquitin conjugating enzyme E2 C; ubiquitin conjugating enzyme E2C; ubiquitin-conjugating enzyme E2 C; ubiquitin-conjugating enzyme E2C; ubiquitin-protein ligase C
Gene Symbols UBE2C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-179 of human UBE2C (NP_0089501).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11065, 296368, 68612
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.