Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2C Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8357R

Description
This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is required for the destruction of mitotic cyclins and for cell cycle progression, and may be involved in cancer progression. Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes ′Lys-11′- and ′Lys-48′-linked polyubiquitination. Acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated ubiquitin ligase that controls progression through mitosis. Acts by initiating ′Lys-11′-linked polyubiquitin chains on APC/C substrates, leading to the degradation of APC/C substrates by the proteasome and promoting mitotic exit.Specifications
UBE2C | |
Polyclonal | |
Unconjugated | |
UBE2C | |
(E3-independent) E2 ubiquitin-conjugating enzyme C; 1110015A16Rik; cyclin-selective ubiquitin carrier protein; D2Ertd695e; dJ447F3.2; E2 ubiquitin-conjugating enzyme C; mitotic-specific ubiquitin-conjugating enzyme; RP3-447F3.1; Ubch10; Ube2c; Ubiquitin carrier protein C; ubiquitin conjugating enzyme E2 C; ubiquitin conjugating enzyme E2C; Ubiquitin-conjugating enzyme E2 C; ubiquitin-conjugating enzyme E2C; Ubiquitin-protein ligase C | |
Rabbit | |
Protein A | |
RUO | |
11065 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O00762 | |
UBE2C | |
KLH conjugated synthetic peptide derived from human UBE2C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction