Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102532 Supplier Thermo Scientific Supplier No. PA5102532
Add to Cart
Add to Cart

UBE2D1 Polyclonal Antibody

UBE2D1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2D1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UBE2D1
Gene Accession No. D3ZDK2, P51668, P61080
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme D1; E2 ubiquitin-conjugating enzyme D1; E2(17)KB 1; E2(17)KB1; SFT; Stimulator of Fe transport; UBC 4/5; UBC4/5; UBC4/5 homolog; UBC5A; UBCH 5; UBCH 5A; UBCH5; UBCH5A; UBE2D1; ubiquitin; Ubiquitin carrier protein D1; ubiquitin conjugating enzyme E2 D1; ubiquitin conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2 D1; Ubiquitin-conjugating enzyme E2(17)KB 1; ubiquitin-conjugating enzyme E2-17 kDa 1; ubiquitin-conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 1, UBC4/5 homolog; ubiquitin-protein ligase D1
Gene Symbols UBE2D1
Host Species Rabbit
Immunogen A synthesized peptide derived from human UBE2D1(Accession P51668), corresponding to amino acid residues V102-M147.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216080, 361831, 7321
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.