Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596006 Supplier Invitrogen™ Supplier No. PA596006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MALKRIQKEL SDLQRDPPAH CSAGPVGDDL FHWQATIMGP PDSAYQGGVF FLTVHFPTDY PFKPPKIAFT TKIYHPNINS NGSICLDILR SQWSPALTVS KVLLSICSLL CDPNPDDPLV PDIAQIYKSD KEKYNRHARE WTQKYAM; Positive Samples: SW620, BT-474; Cellular Location: Cytoplasm.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is closely related to a stimulator of iron transport (SFT), and is up-regulated in hereditary hemochromatosis. It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2D1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE2D1
Gene Accession No. D3ZDK2, P51668, P61080
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme D1; E2 ubiquitin-conjugating enzyme D1; E2(17)KB 1; E2(17)KB1; SFT; Stimulator of Fe transport; UBC 4/5; UBC4/5; UBC4/5 homolog; UBC5A; UBCH 5; UBCH 5A; UBCH5; UBCH5A; UBE2D1; ubiquitin; Ubiquitin carrier protein D1; ubiquitin conjugating enzyme E2 D1; ubiquitin conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2 D1; Ubiquitin-conjugating enzyme E2(17)KB 1; ubiquitin-conjugating enzyme E2-17 kDa 1; ubiquitin-conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 1, UBC4/5 homolog; ubiquitin-protein ligase D1
Gene Symbols UBE2D1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human UBE2D1 (NP_0033291).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216080, 361831, 7321
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.