Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2D2 Monoclonal Antibody (OTI1D11)

UBE2D2 Monoclonal Antibody (OTI1D11)

Supplier:  Thermo Scientific MA526805

Catalog No. PIMA526805


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

UBE2D2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P62837
UBE2D2
Full length human recombinant protein of UBE2D2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1D11
Unconjugated
UBE2D2
(E3-independent) E2 ubiquitin-conjugating enzyme D2; 1500034D03Rik; E2 ubiquitin conjugating enzyme UbcH5B; E2 ubiquitin-conjugating enzyme D2; E2(17)KB 2; E2(17)KB2; E217kB; p53-regulated ubiquitin-conjugating enzyme 1; PUBC1; similar to ubiquitin-conjugating enzyme E2D 2; Ubc2e; ubc2e ubiquitin conjugating enzyme; ubc4; UBC4/5; UBC5B; UBCH4; UBCH5B; ube2d2; Ube2d2a; Ubiquitin carrier protein D2; ubiquitin conjugating enzyme 2e; ubiquitin conjugating enzyme E2 D2; ubiquitin conjugating enzyme E2D; ubiquitin conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2(17)KB 2; ubiquitin-conjugating enzyme E2-17 kDa 2; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 2A; ubiquitin-protein ligase D2; Unknown (protein for MGC:138112); zgc:55886; zgc:55886 protein; zgc:77149; zmp:0000000618
Mouse
Affinity Chromatography
RUO
7322
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.