Learn More
UBE2D2 Mouse anti-Human, Clone: OTI3H1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806599

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.Specifications
UBE2D2 | |
Monoclonal | |
Unconjugated | |
UBE2D2 | |
E2(17)KB2, PUBC1, UBC4, UBC4/5, UBCH4, UBCH5B | |
Mouse | |
Affinity Chromatography | |
RUO | |
7322 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3H1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P62837 | |
UBE2D2 | |
Full length human recombit protein of human UBE2D2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.