Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106767
Description
Antibody detects endogenous levels of total UBE2D2.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
Specifications
| UBE2D2 | |
| Polyclonal | |
| Unconjugated | |
| UBE2D2 | |
| (E3-independent) E2 ubiquitin-conjugating enzyme D2; 1500034D03Rik; E2 ubiquitin conjugating enzyme UbcH5B; E2 ubiquitin-conjugating enzyme D2; E2(17)KB 2; E2(17)KB2; E217kB; p53-regulated ubiquitin-conjugating enzyme 1; PUBC1; similar to ubiquitin-conjugating enzyme E2D 2; Ubc2e; ubc2e ubiquitin conjugating enzyme; ubc4; UBC4/5; UBC5B; UBCH4; UBCH5B; ube2d2; Ube2d2a; Ubiquitin carrier protein D2; ubiquitin conjugating enzyme 2e; ubiquitin conjugating enzyme E2 D2; ubiquitin conjugating enzyme E2D; ubiquitin conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2(17)KB 2; ubiquitin-conjugating enzyme E2-17 kDa 2; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 2A; ubiquitin-protein ligase D2; Unknown (protein for MGC:138112); zgc:55886; zgc:55886 protein; zgc:77149; zmp:0000000618 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56550, 641452, 7322 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62837, P62838, P62839 | |
| UBE2D2, Ube2d2a | |
| A synthesized peptide derived from human UBE2D2(Accession P62837), corresponding to amino acid residues I106-M147. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction