Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2D3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542280
Description
Peptide sequence: MALKRINKEL SDLARDPPAQ CSAGPVGDDM FHWQATIMGP NDSPYQGGVF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Yeast: 92%; Zebrafish: 92%.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Specifications
| UBE2D3 | |
| Polyclonal | |
| Unconjugated | |
| UBE2D3 | |
| (E3-independent) E2 ubiquitin-conjugating enzyme D3; 1100001F19Rik; 9430029A22Rik; AA414951; E2 ubiquitin-conjugating enzyme D3; E2(17)KB 3; E2(17)KB3; eff; fb36f02; LOW QUALITY PROTEIN: ubiquitin-conjugating enzyme E2 D3; MGC43926; MGC5416; PAPase; Phosphoarginine phosphatase; PRO2116; similar to phosphoarginine phosphatase; UBC4/5; UBC5C; UBCH5C; Ube2d3; UBE2D3P; Ubiquitin carrier protein D3; ubiquitin conjugating enzyme E2 D3; ubiquitin conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2(17)KB 3; ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); ubiquitin-protein ligase D3; Unknown (protein for MGC:127427); wu:fb36f02; zgc:56340 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7323 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P61077 | |
| UBE2D3 | |
| Synthetic peptide directed towards the N-terminal of human UBE2D3 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction