Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567548
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567548 Supplier Invitrogen™ Supplier No. PA567548
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Rat samples.

The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2D3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene UBE2D3
Gene Accession No. P61077, P61078, P61079
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme D3; 1100001F19Rik; 9430029A22Rik; AA414951; E2 ubiquitin-conjugating enzyme D3; E2(17)KB 3; E2(17)KB3; eff; fb36f02; LOW QUALITY PROTEIN: ubiquitin-conjugating enzyme E2 D3; MGC43926; MGC5416; PAPase; Phosphoarginine phosphatase; PRO2116; similar to phosphoarginine phosphatase; UBC4/5; UBC5C; UBCH5C; Ube2d3; UBE2D3P; Ubiquitin carrier protein D3; ubiquitin conjugating enzyme E2 D3; ubiquitin conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2(17)KB 3; ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); ubiquitin-protein ligase D3; Unknown (protein for MGC:127427); wu:fb36f02; zgc:56340
Gene Symbols UBE2D3
Host Species Rabbit
Immunogen Full length Fusion protein of human UBE2D3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66105, 7323, 81920
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.