Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589959 Supplier Thermo Scientific Supplier No. PA589959
Add to Cart
Add to Cart

UBE2D3 Polyclonal Antibody

UBE2D3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2D3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UBE2D3
Gene Accession No. P61077, P61078, P61079
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme D3; 1100001F19Rik; 9430029A22Rik; AA414951; E2 ubiquitin-conjugating enzyme D3; E2(17)KB 3; E2(17)KB3; eff; fb36f02; LOW QUALITY PROTEIN: ubiquitin-conjugating enzyme E2 D3; MGC43926; MGC5416; PAPase; Phosphoarginine phosphatase; PRO2116; similar to phosphoarginine phosphatase; UBC4/5; UBC5C; UBCH5C; Ube2d3; UBE2D3P; Ubiquitin carrier protein D3; ubiquitin conjugating enzyme E2 D3; ubiquitin conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2(17)KB 3; ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); ubiquitin-protein ligase D3; Unknown (protein for MGC:127427); wu:fb36f02; zgc:56340
Gene Symbols UBE2D3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBE2D3 (NP_0033311).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66105, 7323, 81920
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.