Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2D3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119881
This item is not returnable.
View return policy
Description
Positive Control: Jurkat cell lysate, 293T cell lysate, MCF-7 cell lysate, rat liver tissue lysate, mouse lung tissue lysate, SW620, mouse brain tissue. Subcellular Location: Cell membrane, Endosome membrane.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Specifications
UBE2D3 | |
Polyclonal | |
Unconjugated | |
UBE2D3 | |
(E3-independent) E2 ubiquitin-conjugating enzyme D3; 1100001F19Rik; 9430029A22Rik; AA414951; E2 ubiquitin-conjugating enzyme D3; E2(17)KB 3; E2(17)KB3; eff; fb36f02; LOW QUALITY PROTEIN: ubiquitin-conjugating enzyme E2 D3; MGC43926; MGC5416; PAPase; Phosphoarginine phosphatase; PRO2116; similar to phosphoarginine phosphatase; UBC4/5; UBC5C; UBCH5C; Ube2d3; UBE2D3P; Ubiquitin carrier protein D3; ubiquitin conjugating enzyme E2 D3; ubiquitin conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2 D3; Ubiquitin-conjugating enzyme E2(17)KB 3; ubiquitin-conjugating enzyme E2-17 kDa 3; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 3; ubiquitin-conjugating enzyme E2D 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 3 (UBC4/5 homolog, yeast); ubiquitin-protein ligase D3; Unknown (protein for MGC:127427); wu:fb36f02; zgc:56340 | |
Rabbit | |
Protein A | |
RUO | |
66105, 7323, 81920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P61077, P61078, P61079 | |
UBE2D3 | |
Recombinant full length protein of human UBE2D3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction