Learn More
Invitrogen™ UBE2D4 Monoclonal Antibody (3E1)

Description
Peptide Sequence: MALKRIQKEL TDLQRDPPAQ CSAGPVGDDL FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TKIYHPNINS NGSICLDILR SQWS.
Specifications
Specifications
| Antigen | UBE2D4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3E1 |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Ube2d4 |
| Gene Accession No. | Q9Y2X8 |
| Gene Alias | E2 ubiquitin-conjugating enzyme D2B; E2 ubiquitin-conjugating enzyme D4; E2(17)KB 2B; HBUCE1; RGD69425; UBCH5D; Ube2d2; Ube2d2b; UBE2D4; Ubiquitin carrier protein D2B; ubiquitin carrier protein D4; ubiquitin conjugating enzyme E2 D4 (putative); ubiquitin conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme E2 D2B; Ubiquitin-conjugating enzyme E2 D4; ubiquitin-conjugating enzyme E2(17)KB 2B; Ubiquitin-conjugating enzyme E2-17 kDa 2B; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme HBUCE1; ubiquitin-protein ligase D2B; Ubiquitin-protein ligase D4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.