Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2DNL Monoclonal Antibody (2F7)
GREENER_CHOICE

Catalog No. PIMA523263
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523263 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523263 Supplier Invitrogen™ Supplier No. MA523263
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MALKLIHKEF LELARDPQPH CSAGPVWDDM LHWQATITRP NDSSYLGGVF FLKFPSDYLF KPPKIKFTNG IYHQR.

TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2DNL
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene UBE2DNL
Gene Accession No. Q8IWF7
Gene Alias UBE2DNL; ubiquitin conjugating enzyme E2 D N-terminal like (pseudogene); ZNF553
Gene Symbols UBE2DNL
Host Species Mouse
Immunogen UBE2DNL (AAH40290, 1 a.a. ~ 75 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131816
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.