Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542281 Supplier Invitrogen™ Supplier No. PA542281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PKGDNIYEWR STILGPPGSV YEGGVFFLDI TFTPEYPFKP PKVTFRTRIY Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

UBE2E1 catalyzes the covalent attachment of ubiquitin to other proteins. UBE2E1 mediates the selective degradation of short-lived and abnormal proteins.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2E1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UBE2E1
Gene Accession No. P51965
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme E1; E2 ubiquitin-conjugating enzyme E1; Ubce5; UBCH6; ubcM2; UbcM3; UBE2E1; Ubiquitin carrier protein E1; ubiquitin conjugating enzyme E2 E1; ubiquitin conjugating enzyme E2E 1; ubiquitin-conjugating enzyme 5; ubiquitin-conjugating enzyme E2 E1; Ubiquitin-conjugating enzyme E2 E1-like protein; ubiquitin-conjugating enzyme E2E 1; ubiquitin-conjugating enzyme E2E 1 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 1, UBC4/5 homolog; ubiquitin-protein ligase E1; zgc:171646
Gene Symbols UBE2E1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human UBE2E1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7324
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.