Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2E2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP180573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180573 Supplier Novus Biologicals Supplier No. NBP180573
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2E2 Polyclonal specifically detects UBE2E2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen UBE2E2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_689866.1
Gene Alias EC 6.3.2.19, FLJ25157, UbcH8, Ubiquitin carrier protein E2, ubiquitin-conjugating enzyme E2 E2, ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E2
Gene Symbols UBE2E2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 25 and 75 of human Ubiquitin-conjugating Enzyme E2E 2 using the numbering given in entry NP_689866.1 (GeneID 7325).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7325
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.