Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555996 Supplier Invitrogen™ Supplier No. PA555996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STEAQRVDDS PSTSGGSSDG DQRESVQQEP EREQVQPKKK EGKISSKTAA KLSTSAKRIQ KELAEITLDP PPNCSAGPKG DNIYEWRSTI LGPPGSVYEG GVFFLDITFS PDYPFKPPKV TFRTRIYHC Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 66%.

ubiquitin-conjugating enzyme E2 (UBE2E2) is a ubiquitin conjugating enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2E2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2E2
Gene Accession No. Q91W82, Q96LR5
Gene Alias BC016265; E2 ubiquitin-conjugating enzyme E2; FLJ25157; TBC1 domain family, member 12; UB2E2; UBCH8; UBE2E2; Ubiquitin carrier protein E2; ubiquitin conjugating enzyme E2 E2; ubiquitin conjugating enzyme E2E 2; ubiquitin-conjugating enzyme E2 E2; ubiquitin-conjugating enzyme E2E 2; ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast); ubiquitin-protein ligase E2; wu:fb38h07; zgc:92467
Gene Symbols UBE2E2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBE2E2. Recombinant protein control fragment (Product #RP-102080).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218793, 361013, 7325
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.