Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2E2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565263
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5263 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5263 Supplier Bethyl Laboratories, Inc Supplier No. A303485AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.

Specifications

Antigen UBE2E2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene UBE2E2
Gene Accession No. Q96LR5
Gene Alias E2 ubiquitin-conjugating enzyme E2, FLJ25157, UbcH8, ubiquitin carrier protein E2, ubiquitin conjugating enzyme E2E 2, ubiquitin-conjugating enzyme E2E 2, ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast), ubiquitin-protein ligase E2
Gene Symbols UBE2E2
Host Species Rabbit
Immunogen Between 25 and 75
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7325
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.